Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163430.1 | NP_001156902.1 | 4 | 764 | ||
NM_181586.3 | NP_853617.1 | 4 | 470 | |||
GenBank mRNA | AK013316.1 | - | 4 | 419 | ||
AK145929.1 | - | 4 | 764 | |||
BC052763.1 | - | 4 | 420 | 6513375 |