Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icosl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015790.3 | NP_056605.1 | 4 | 837 | ||
GenBank mRNA | AF199027.1 | - | 4 | 746 | ||
AF216747.1 | - | 3 | 680 | |||
AF394451.1 | - | 680 | ||||
AK041320.1 | - | 3 | 1305 | |||
AK041421.1 | - | 4 | 847 | |||
AK136074.1 | - | 1 | 19 | |||
AK220220.1 | - | 1 | 564 | |||
BC029227.1 | - | 4 | 837 | 4217333 |