Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015791.3 | NP_056606.2 | 2 | 433 | ||
GenBank mRNA | AF176527.1 | - | 2 | 440 | ||
AK002538.1 | - | 2 | 444 | |||
AK012401.1 | - | 2 | 451 | |||
AK029989.1 | - | 2 | 434 | |||
AK077595.1 | - | 2 | 455 | |||
AK078111.1 | - | 2 | 453 | |||
AK147748.1 | - | 2 | 450 | |||
AK160145.1 | - | 2 | 414 | |||
AK162592.1 | - | 2 | 450 | |||
BC011100.1 | - | 2 | 454 | 4011855 | ||
BC034854.1 | - | 2 | 365 | 3468704 |