Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015791.3 | NP_056606.2 | 2 | 408 | ||
GenBank mRNA | AF176527.1 | - | 2 | 415 | ||
AK002538.1 | - | 2 | 419 | |||
AK012401.1 | - | 2 | 426 | |||
AK029989.1 | - | 2 | 409 | |||
AK077595.1 | - | 2 | 430 | |||
AK078111.1 | - | 2 | 428 | |||
AK147748.1 | - | 2 | 425 | |||
AK160145.1 | - | 2 | 389 | |||
AK162592.1 | - | 2 | 425 | |||
BC011100.1 | - | 2 | 429 | 4011855 | ||
BC034854.1 | - | 2 | 340 | 3468704 |