Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015793.2 | NP_056608.1 | 951 | |||
GenBank mRNA | AK087709.1 | - | 951 | |||
AK135682.1 | - | 947 | ||||
AK166611.1 | - | 789 | ||||
AK166612.1 | - | 789 | ||||
BC099372.1 | - | 776 | 30938910 | |||
BC138018.1 | - | 945 | 8861037 | |||
BC145098.1 | - | 782 | 9053646 |