Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282061.1 | NP_001268990.1 | 9 | 638 | ||
NM_015812.3 | NP_056627.1 | 9 | 920 | |||
GenBank mRNA | AF061933.1 | - | 6 | 365 | ||
AK052527.1 | - | 9 | 921 | |||
AK053087.1 | - | 9 | 639 | |||
AK139323.1 | - | 9 | 1035 | |||
BC038616.1 | - | 7 | 465 | |||
BC125029.1 | - | 9 | 624 | 40062355 | ||
BC125030.1 | - | 9 | 624 | 40062356 |