Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081650.2 | NP_001075119.1 | 10 | 1145 | ||
NM_019492.3 | NP_062365.2 | 19 | 1849 | |||
NM_134257.3 | NP_599018.3 | 10 | 1145 | |||
GenBank mRNA | AF319519.1 | - | 10 | 1158 | ||
AF350047.1 | - | 10 | 1128 | |||
AJ250999.1 | - | 19 | 1849 | |||
AK004648.1 | - | 4 | 314 | |||
AK015722.1 | - | 17 | 1606 | |||
AK028445.1 | - | 10 | 1130 | |||
AK041392.1 | - | 4 | 321 | |||
AK046265.1 | - | 1 | 158 | |||
BC131680.1 | - | 1 | 266 | |||
BY704855.1 | - | 4 | 309 |