Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081650.2 | NP_001075119.1 | 8, 9 | 1097 | ||
NM_019492.3 | NP_062365.2 | 17, 18 | 1801 | |||
NM_134257.3 | NP_599018.3 | 8, 9 | 1097 | |||
GenBank mRNA | AF319519.1 | - | 8, 9 | 1110 | ||
AF350047.1 | - | 8, 9 | 1080 | |||
AJ250999.1 | - | 17, 18 | 1801 | |||
AK004648.1 | - | 2, 3 | 266 | |||
AK015722.1 | - | 15, 16 | 1558 | |||
AK028445.1 | - | 8, 9 | 1082 | |||
AK041392.1 | - | 2, 3 | 273 | |||
BY704855.1 | - | 2, 3 | 261 |