Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081650.2 | NP_001075119.1 | 6 | 717 | ||
NM_019492.3 | NP_062365.2 | 15 | 1421 | |||
NM_134257.3 | NP_599018.3 | 6 | 717 | |||
GenBank mRNA | AF319519.1 | - | 6 | 730 | ||
AF350047.1 | - | 6 | 700 | |||
AJ250999.1 | - | 15 | 1421 | |||
AK015722.1 | - | 13 | 1176 | |||
AK028445.1 | - | 6 | 702 |