Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015820.3 | NP_056635.2 | 2 | 894 | ||
GenBank mRNA | AB024567.1 | - | 2 | 857 | ||
BC117529.1 | - | 3 | 712 | 40106840 | ||
BC118035.1 | - | 2 | 818 | 40106835 |