Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 7 | 888 | ||
GenBank mRNA | AF285434.1 | - | 6 | 819 | ||
AY366242.1 | - | 7 | 888 | |||
KP260924.1 | - | 7 | 855 | |||
U24075.1 | - | 7 | 873 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF285432.1 | - | 6 | 819 |