Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015822.2 | NP_056637.1 | 5 | 1261 | ||
GenBank mRNA | AF176521.1 | - | 5 | 968 | ||
AK080802.1 | - | 6 | 1390 | |||
AK080863.1 | - | 5 | 1270 | |||
AK142007.1 | - | 5 | 1261 | |||
AK166128.1 | - | 5 | 1143 | |||
AK167409.1 | - | 5 | 1080 | |||
BC026835.1 | - | 4 | 928 | |||
BC037665.1 | - | 4 | 924 | |||
BC046330.1 | - | 5 | 975 | |||
BC067203.1 | - | 5 | 1143 | 6334317 |