Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190414.1 | NP_001177343.1 | 5 | 1135 | ||
NM_015828.3 | NP_056643.3 | 5 | 953 | |||
GenBank mRNA | AJ132236.1 | - | 4 | 781 | ||
AK010070.1 | - | 1, 2 | 101 | |||
AK136573.1 | - | 5 | 954 | |||
AK170707.1 | - | 5 | 1136 | |||
AK191922.1 | - | 351 | ||||
AK197576.1 | - | 3 | 209 | |||
AK219474.1 | - | 3 | 209 | |||
AM697710.1 | - | 5 | 874 | |||
BC015277.1 | - | 5 | 941 | 4216299 | ||
BC051254.1 | - | 5 | 941 | 6478532 |