Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177572.1 | NP_001171043.1 | 18 | 2108 | ||
NM_015829.3 | NP_056644.1 | 18 | 2111 | |||
GenBank mRNA | AF164632.1 | - | 18 | 2056 | ||
AK002829.1 | - | 18 | 1986 | |||
AK012670.1 | - | 18 | 1999 | |||
AK029020.1 | - | 18 | 2039 | |||
AK150496.1 | - | 18 | 2053 | |||
AK151180.1 | - | 18 | 2053 | |||
AK152237.1 | - | 18 | 2053 | |||
BC016571.1 | - | 12 | 1245 | 3600909 |