Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016698.2 | NP_057907.2 | 7, 8 | 1609 | ||
GenBank mRNA | AB026621.1 | - | 7, 8 | 1608 | ||
AK004579.1 | - | 7, 8 | 1604 | |||
AK034953.1 | - | 7, 8 | 1608 | |||
AK129106.1 | - | 7, 8 | 1597 | |||
AK146728.1 | - | 7, 8 | 1609 | |||
AK149972.1 | - | 7, 8 | 1610 | |||
AK156502.1 | - | 7, 8 | 1606 | |||
AK165617.1 | - | 7, 8 | 1609 | |||
AK168523.1 | - | 7, 8 | 1604 | |||
AK172065.1 | - | 7, 8 | 1607 | |||
AK182623.1 | - | 1, 2 | 101 | |||
AK215782.1 | - | 2, 3 | 189 | |||
AK218316.1 | - | 1, 2 | 101 | |||
BC010342.1 | - | 7, 8 | 1318 | 3602969 |