Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080770.1 | NP_001074239.1 | 7 | 902 | ||
NM_001080772.1 | NP_001074241.1 | 8 | 1006 | |||
NM_002255.5 | NP_002246.5 | 8 | 1007 | |||
GenBank mRNA | AF002256.1 | - | 6 | 573 | ||
AF002979.1 | - | 8 | 967 | |||
AF002980.1 | - | 7 | 874 | |||
AF002981.1 | - | 6 | 823 | |||
AF002982.1 | - | 5 | 769 | |||
AF034771.1 | - | 8 | 973 | |||
AF034772.1 | - | 8 | 973 | |||
AF034773.1 | - | 8 | 973 | |||
AF276292.1 | - | 8 | 966 | |||
AF285436.1 | - | 8 | 994 | |||
AY223512.2 | - | 6 | 893 | |||
AY223513.1 | - | 6 | 894 | |||
AY223514.1 | - | 6 | 893 | |||
AY223515.1 | - | 5 | 789 | |||
AY250088.1 | - | 6 | 894 | |||
AY359817.1 | - | 8 | 978 | |||
AY601812.1 | - | 8 | 1027 | |||
AY789058.1 | - | 8 | 961 | |||
AY789059.1 | - | 8 | 961 | |||
AY789060.1 | - | 8 | 960 | |||
AY789061.1 | - | 8 | 960 | |||
AY789062.1 | - | 8 | 961 | |||
BC041611.1 | - | 8 | 1008 | 5485491 | ||
DQ266438.1 | - | 8 | 970 | |||
EU194340.1 | - | 8 | 967 | |||
EU194341.1 | - | 6 | 766 | |||
EU194342.1 | - | 8 | 966 | |||
U71199.1 | - | 8 | 979 | |||
U73394.1 | - | 7 | 928 | |||
X97229.1 | - | 8 | 972 | |||
X99479.1 | - | 7 | 916 | |||
X99480.1 | - | 8 | 978 | |||
Y13054.1 | - | 7 | 862 |