Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290745.1 | NP_001277674.1 | 26 | 2960 | ||
NM_016965.3 | NP_058661.1 | 25 | 2942 | |||
GenBank mRNA | AJ534525.1 | - | 25 | 2725 | ||
AK122318.1 | - | 24 | 2763 | |||
AK143049.1 | - | 2 | 100 | |||
BC003962.1 | - | 5 | 464 | |||
BC032206.1 | - | 4 | 304 | |||
BC060197.1 | - | 25 | 2920 | |||
BC127067.1 | - | 26 | 2742 | |||
X61453.1 | - | 17 | 1864 |