Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016710.2 | NP_057919.2 | 4 | 208 | ||
GenBank mRNA | AB018374.2 | - | 4 | 163 | ||
AF213454.1 | - | 4 | 188 | |||
AK013748.1 | - | 3 | 77 | |||
AK131996.1 | - | 4 | 178 | |||
AK146755.1 | - | 4 | 184 | |||
BC021626.1 | - | 4 | 160 | 4219308 | ||
BC083087.1 | - | 4 | 170 | 6518798 | ||
BY279928.1 | - | 4 | 233 |