Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019583.3 | NP_062529.2 | 7 | 659 | ||
GenBank mRNA | AF208108.1 | - | 7 | 572 | ||
AF208109.1 | - | 7 | 1198 | |||
AK045627.1 | - | 7 | 660 | |||
AK050139.1 | - | 7 | 580 | |||
BC026546.1 | - | 7 | 559 | 5042466 |