Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097617.1 | NP_001091086.1 | 12 | 1819 | ||
NM_144938.2 | NP_659187.2 | 12 | 1856 | |||
GenBank mRNA | AF459019.1 | - | 12 | 1783 | ||
AK087522.1 | - | 12 | 1788 | |||
AK172453.1 | - | 12 | 1784 | |||
BC018319.1 | - | 12 | 1748 | 4196654 | ||
BC022123.1 | - | 12 | 1809 | 5097206 | ||
BC111880.1 | - | 1612 | 40059749 |
Gene Symbol: C1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173864.2 | NP_776289.2 | 11 | 1630 | ||
GenBank mRNA | AF459020.1 | - | 11 | 1630 |