Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016699.2 | NP_057908.2 | 3 | 396 | ||
GenBank mRNA | AF091392.1 | - | 3 | 365 | ||
AK017621.1 | - | 3 | 369 | |||
AK081014.1 | - | 3 | 392 | |||
AK144162.1 | - | 3 | 381 | |||
AK162887.1 | - | 3 | 365 | |||
AK166070.1 | - | 3 | 380 | |||
AK168700.1 | - | 3 | 369 | |||
BC027326.1 | - | 3 | 374 | 3987812 | ||
BC027840.1 | - | 3 | 384 | 5325071 |