Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016719.1 | NP_057928.1 | 6 | 819 | ||
GenBank mRNA | AF155647.1 | - | 6 | 819 | ||
AK010849.1 | - | 6 | 836 | |||
AK010903.1 | - | 6 | 836 | |||
AK034134.1 | - | 1 | 1110 | |||
AK147041.1 | - | 6 | 828 | |||
BC138581.1 | - | 6 | 725 | 8861601 | ||
BC145913.1 | - | 6 | 725 | 40131216 |