Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ249491.1 | - | 3 | 317 |
Gene Symbol: Iltifb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054079.2 | NP_473420.2 | 3 | 313 |