Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045959.1 | NP_001039424.1 | 22 | 2803 | ||
NM_001045964.1 | NP_001039429.1 | 23 | 2824 | |||
NM_016713.2 | NP_057922.2 | 21 | 2692 | |||
NM_176893.2 | NP_795712.2 | 22 | 2716 | |||
GenBank mRNA | AK220178.1 | - | 4 | 242 | ||
BC011346.1 | - | 18 | 2196 | |||
BC052474.1 | - | 22 | 2658 | 6833893 | ||
BC079549.1 | - | 21 | 3174 | |||
BU707794.1 | - | 7 | 533 | |||
CF736764.1 | - | 8 | 706 |