Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016750.3 | NP_058030.1 | 4 | 482 | ||
GenBank mRNA | AK028176.1 | - | 4 | 379 | ||
AK088704.1 | - | 4 | 429 | |||
AK143572.1 | - | 4 | 381 | |||
AK146181.1 | - | 4 | 414 | |||
AK150454.1 | - | 4 | 381 | |||
AK151052.1 | - | 4 | 381 | |||
AK151462.1 | - | 3, 4 | 310 | |||
AK151509.1 | - | 4 | 381 | |||
AK151868.1 | - | 4 | 381 | |||
AK152398.1 | - | 4 | 379 | |||
AK152848.1 | - | 4 | 380 | |||
AK168178.1 | - | 4 | 381 | |||
AK169003.1 | - | 4 | 267 | |||
AY074806.1 | - | 4 | 379 | |||
BC079903.1 | - | 4 | 404 | 30622912 | ||
BC147478.1 | - | 4 | 319 | 9056200 | ||
BC158035.1 | - | 4 | 319 | 9007338 | ||
BC171999.1 | - | 4 | 319 | 9054285 | ||
U70494.1 | - | 4 | 370 |