Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016758.3 | NP_058038.2 | 4 | 444 | ||
GenBank mRNA | AK154944.1 | - | 4 | 369 | ||
BC030321.1 | - | 4 | 416 | 5374531 | ||
U85055.1 | - | 4 | 284 |