Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164156.1 | NP_001157628.1 | 7 | 862 | ||
NM_016766.3 | NP_058046.2 | 8 | 982 | |||
GenBank mRNA | AF015309.1 | - | 8 | 932 | ||
AK088116.1 | - | 8 | 947 | |||
AK158347.1 | - | 8 | 962 | |||
AK158611.1 | - | 4 | 589 | |||
AK167242.1 | - | 7 | 822 | |||
AK181698.1 | - | 3 | 280 | |||
AK184558.1 | - | 3 | 280 | |||
AK184749.1 | - | 3 | 280 | |||
AK209104.1 | - | 1 | 250 | |||
BC003746.1 | - | 8 | 966 | 3486681 | ||
BC085099.1 | - | 8 | 950 | 6391692 | ||
BC108340.1 | - | 8 | 957 | 4504579 |