Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122982.2 | NP_001116454.1 | 8, 9 | 724 | ||
NM_001290420.1 | NP_001277349.1 | 8, 9 | 724 | |||
NM_001290422.1 | NP_001277351.1 | 8, 9 | 724 | |||
NM_016746.4 | NP_058026.2 | 8, 9 | 724 | |||
GenBank mRNA | AK009615.1 | - | 8, 9 | 670 | ||
AK014079.1 | - | 7, 8 | 573 | |||
AK037957.1 | - | 8, 9 | 725 | |||
AK135472.1 | - | 8, 9 | 720 | |||
AK153035.1 | - | 8, 9 | 657 | |||
BC003344.1 | - | 8, 9 | 657 | 3257730 | ||
BC029211.1 | - | 8, 9 | 666 | 4526925 | ||
BC037689.1 | - | 8, 9 | 652 | |||
BC062376.1 | - | 8, 9 | 639 | |||
BC087544.1 | - | 8, 9 | 656 | |||
BC100396.1 | - | 8, 9 | 698 | |||
BC120649.1 | - | 8, 9 | 651 | 40129572 | ||
BC120677.1 | - | 8, 9 | 651 | 40129600 | ||
U62638.1 | - | 8, 9 | 699 |