Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290824.1 | NP_001277753.1 | 15 | 1860 | ||
NM_153387.3 | NP_700436.1 | 15 | 1863 | |||
GenBank mRNA | AK034694.1 | - | 6 | 1391 | ||
AK038921.1 | - | 15 | 1819 | |||
AK045305.1 | - | 15 | 1756 | |||
AK046132.1 | - | 15 | 1793 | |||
AK050250.1 | - | 15 | 1861 | |||
AK051517.1 | - | 15 | 1761 | |||
AK152984.1 | - | 15 | 1819 | |||
AK153171.1 | - | 15 | 1819 | |||
AK161774.1 | - | 6 | 659 | |||
AK179523.1 | - | 3 | 294 | |||
AK185696.1 | - | 2 | 105 | |||
AK214198.1 | - | 3 | 294 | |||
BC029106.1 | - | 15 | 1827 | 3711922 |