Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159864.1 | NP_001153336.1 | 4 | 561 | ||
GenBank mRNA | AK032658.1 | - | 4 | 530 | ||
AK132699.1 | - | 4 | 530 | |||
BC075692.1 | - | 4 | 593 | 30631161 | ||
BY736102.1 | - | 4 | 533 |