Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159864.1 | NP_001153336.1 | 3 | 475 | ||
GenBank mRNA | AK032658.1 | - | 3 | 444 | ||
AK053098.1 | - | 2 | 2427 | |||
AK079700.1 | - | 3 | 471 | |||
AK132699.1 | - | 3 | 444 | |||
BY736102.1 | - | 3 | 447 |