Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026373.4 | NP_080649.1 | 4 | 412 | ||
GenBank mRNA | AK003686.2 | - | 4 | 378 | ||
AK012121.1 | - | 4 | 415 | |||
AK018035.1 | - | 4 | 412 | |||
BC025656.1 | - | 4 | 378 | 5342719 |