Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198937.2 | NP_945175.1 | 2 | 180 | ||
GenBank mRNA | AK150226.1 | - | 2 | 192 | ||
AK152133.1 | - | 2 | 196 | |||
AK152573.1 | - | 2 | 194 | |||
AK166213.1 | - | 2 | 194 | |||
AK167223.1 | - | 2 | 164 | |||
AK169647.1 | - | 227 | ||||
AK180614.1 | - | 2 | 154 | |||
AK212681.1 | - | 1 | 110 | |||
BC057024.1 | - | 2 | 180 | 6815840 | ||
BC080740.1 | - | 2 | 220 | 30460749 |