Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177465.4 | NP_803416.2 | 13 | 2538 | ||
GenBank mRNA | AK040446.1 | - | 5 | 1039 | ||
AY771616.1 | - | 14 | 2455 | |||
AY771617.1 | - | 13 | 2542 | |||
AY771618.1 | - | 13 | 2197 | |||
BC051427.1 | - | 13 | 2193 |