Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029320.3 | NP_083596.2 | 2, 3 | 972 | ||
GenBank mRNA | AK015781.1 | - | 2, 3 | 439 | ||
AK017107.1 | - | 2, 3 | 973 | |||
AK030385.1 | - | 2, 3 | 453 | |||
AK045133.1 | - | 2, 3 | 467 | |||
AL361753.1 | - | 1, 2 | 103 | |||
BC113804.1 | - | 2, 3 | 449 | |||
BC129811.1 | - | 2, 3 | 449 | 40062382 | ||
BC129812.1 | - | 2, 3 | 449 | 40062383 |