Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033128.3 | NP_001028300.1 | 4 | 365 | ||
GenBank mRNA | AK037753.1 | - | 4 | 351 | ||
AK045850.1 | - | 4 | 363 | |||
AK161343.1 | - | 4 | 359 | |||
AK168901.1 | - | 4 | 366 | |||
BC076577.1 | - | 4 | 354 |