Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003955.2 | NP_001003955.1 | 1 | 265 | ||
NM_177466.4 | NP_803417.3 | 1 | 265 | |||
GenBank mRNA | AK129230.1 | - | 1 | 265 | ||
AK154922.1 | - | 1 | 248 | |||
BC141380.1 | - | 1 | 195 |