Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003955.2 | NP_001003955.1 | 2 | 608 | ||
NM_177466.4 | NP_803417.3 | 2 | 608 | |||
GenBank mRNA | AK129230.1 | - | 2 | 608 | ||
AK145061.1 | - | 2 | 121 | |||
AK154922.1 | - | 2 | 591 | |||
BC044833.1 | - | 2 | 137 | |||
BC051063.1 | - | 2 | 137 | |||
BC141380.1 | - | 2 | 538 |