Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026504.2 | NP_080780.1 | 6, 7 | 885 | ||
GenBank mRNA | AK007496.1 | - | 6, 7 | 886 | ||
AK014348.1 | - | 6, 7 | 897 | |||
AK045993.1 | - | 6, 7 | 892 | |||
AK050334.1 | - | 6, 7 | 892 | |||
AK216563.1 | - | 2, 3 | 154 | |||
BC052345.1 | - | 6, 7 | 892 | 6585618 |