Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081279.1 | NP_001074748.1 | 1 | 2388 | ||
GenBank mRNA | AK132349.1 | - | 1 | 1597 | ||
AK171731.1 | - | 1 | 1597 | |||
AK171759.1 | - | 1 | 1230 | |||
BC082308.1 | - | 1 | 2388 |