Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgsnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029884.1 | NP_084160.1 | 8 | 863 | ||
GenBank mRNA | AK035264.1 | - | 8 | 796 | ||
AK149883.1 | - | 8 | 867 | |||
AK152926.1 | - | 8 | 828 | |||
AK159649.1 | - | 8 | 792 | |||
AK160068.1 | - | 8 | 786 | |||
AK209791.1 | - | 2 | 129 | |||
BC024084.1 | - | 8 | 770 | 5101064 |