Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028771.2 | NP_083047.1 | 2, 3 | 594 | ||
GenBank mRNA | AK004762.1 | - | 2, 3 | 595 | ||
AK013304.1 | - | 2, 3 | 545 | |||
AK045509.1 | - | 2, 3 | 586 | |||
AK088221.1 | - | 2, 3 | 585 | |||
AK155466.1 | - | 2, 3 | 577 | |||
BC006752.1 | - | 1, 2 | 99 | |||
BC078634.1 | - | 2, 3 | 561 | 5388281 |