Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002231.2 | NP_001002231.1 | 3 | 370 | ||
NM_001256080.1 | NP_001243009.1 | 2 | 210 | |||
NM_005551.4 | NP_005542.1 | 3 | 370 | |||
NR_045762.1 | 3 | 370 | ||||
NR_045763.1 | 3 | 456 | ||||
GenBank mRNA | AF188745.1 | - | 3 | 345 | ||
AF188746.1 | - | 3 | 345 | |||
AF188747.1 | - | 3 | 345 | |||
AK300527.1 | - | 3 | 456 | |||
AK300549.1 | - | 2 | 204 | |||
AK300566.1 | - | 3 | 364 | |||
AK309968.1 | - | 1 | 345 | |||
AK309969.1 | - | 3 | 309 | |||
AK309973.1 | - | 2 | 204 | |||
AK309982.1 | - | 2 | 210 | |||
AK309985.1 | - | 2 | 207 | |||
AK315890.1 | - | 2 | 197 | |||
AY429509.1 | - | 3 | 358 | |||
AY429510.1 | - | 2 | 198 | |||
BC005196.1 | - | 3 | 370 | 3951036 | ||
BT006650.1 | - | 3 | 329 | |||
DA865689.1 | - | 3 | 367 | |||
S39329.1 | - | 3 | 345 |