Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162538.1 | NP_001156010.1 | 4 | 542 | ||
NM_025714.4 | NP_079990.2 | 4 | 408 | |||
GenBank mRNA | AK014630.1 | - | 4 | 410 | ||
AK016726.1 | - | 4 | 524 | |||
AK031564.1 | - | 4 | 544 | |||
AK048931.1 | - | 3 | 1321 | |||
AK085115.1 | - | 4 | 532 | |||
BB621104.1 | - | 4 | 544 |