Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162538.1 | NP_001156010.1 | 9 | 1105 | ||
NM_001162539.1 | NP_001156011.1 | 8 | 976 | |||
NM_025714.4 | NP_079990.2 | 9 | 971 | |||
GenBank mRNA | AK014630.1 | - | 9 | 973 | ||
AK016726.1 | - | 9 | 1087 | |||
AK031564.1 | - | 9 | 1107 | |||
AK048931.1 | - | 7 | 2563 | |||
AK085115.1 | - | 9 | 1097 | |||
BC020075.1 | - | 8 | 945 | 3979963 |