Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172762.2 | NP_766350.2 | 4 | 434 | ||
GenBank mRNA | AK016057.1 | - | 4 | 426 | ||
AK078099.1 | - | 4 | 427 | |||
AK079563.1 | - | 4 | 405 | |||
AK136464.1 | - | 4 | 405 | |||
BC064035.1 | - | 4 | 413 | |||
BC141278.1 | - | 4 | 423 | 9055890 |