Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172762.2 | NP_766350.2 | 11 | 1220 | ||
GenBank mRNA | AK011158.1 | - | 2 | 183 | ||
AK016057.1 | - | 10 | 1152 | |||
AK078099.1 | - | 11 | 1214 | |||
AK218432.1 | - | 1 | 21 | |||
AK220250.1 | - | 2 | 221 | |||
BC064035.1 | - | 11 | 1199 | |||
BC141278.1 | - | 11 | 1209 | 9055890 |