Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024213.2 | NP_077175.1 | 21 | 1673 | ||
GenBank mRNA | AK011994.1 | - | 4 | 232 | ||
AK088759.1 | - | 21 | 1663 | |||
AK146000.1 | - | 21 | 1670 | |||
AK148625.1 | - | 21 | 1683 | |||
AK166061.1 | - | 21 | 1796 | |||
AK168897.1 | - | 22 | 1755 | |||
BC016237.1 | - | 21 | 1656 | 4504779 | ||
BC024870.1 | - | 21 | 1673 | 4949079 |