Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033560.3 | NP_291038.2 | 10 | 1355 | ||
GenBank mRNA | AK016385.1 | - | 10 | 1338 | ||
AK157507.1 | - | 10 | 1322 | |||
AK171055.1 | - | 10 | 1344 | |||
AK218075.1 | - | 5 | 356 | |||
BC039290.1 | - | 10 | 1311 | 5066508 | ||
BC110396.1 | - | 10 | 1270 | 5253683 | ||
BY549007.1 | - | 1 | 12 |