Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026555.2 | NP_080831.2 | 6 | 880 | ||
GenBank mRNA | AK003918.1 | - | 5 | 550 | ||
AK077943.1 | - | 6 | 820 | |||
AK082762.1 | - | 6 | 810 | |||
AK086705.1 | - | 6 | 821 | |||
BC005487.1 | - | 5 | 717 | |||
BC025602.1 | - | 6 | 831 | 5250505 | ||
BC055903.1 | - | 6 | 827 | 4010685 | ||
BC092069.1 | - | 6 | 778 | 4215821 |